ENA428074348 Enamine

(1R)-1-(pyridin-2-yl)propan-1-ol

  • MDL number MFCD20527980
  • Cas Number : 18209-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181731
Availability details for
Quantity
  • Availability
  • Add to Cart