ENA428074353 Enamine

2-methyl-4-sulfanylbutan-2-ol

  • MDL number MFCD28064004
  • Cas Number : 33959-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-181777

Safety Information

signalword Warning
pcodes P210,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P370+P378,P403+P233,P403+P235,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-181777
pictograms GHS07
hstatements H227,H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart