ENA428074361 Enamine

5-bromo-6-fluoro-2,3-dihydro-1-benzofuran-3-one

  • MDL number MFCD18208077
  • Cas Number : 1260007-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-181823
Availability details for
Quantity
  • Availability
  • Add to Cart