ENA428074368 Enamine

(3-amino-2-phenylpropyl)dimethylamine

  • MDL number MFCD12192888
  • Cas Number : 259537-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178914
Availability details for
Quantity
  • Availability
  • Add to Cart