ENA428074380 Enamine

5-chloro-N-methyl-1,3-benzothiazol-2-amine

  • MDL number MFCD04111218
  • Cas Number : 34551-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181879
Availability details for
Quantity
  • Availability
  • Add to Cart