ENA428074382 Enamine

2-(chloromethoxy)-1,1,1-trifluoroethane

  • MDL number MFCD19232647
  • Cas Number : 21937-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181881
Availability details for
Quantity
  • Availability
  • Add to Cart