ENA428074383 Enamine

(1-cyclopropylpiperidin-2-yl)methanamine

  • MDL number MFCD26636504
  • Cas Number : 1515351-76-4
  • Molecular Weight : 154.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-181882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181882
Availability details for
Quantity
  • Availability
  • Add to Cart