ENA428074385 Enamine

4-cyclobutyl-1,3-thiazol-2-amine

  • MDL number MFCD14706925
  • Cas Number : 475058-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181894
Availability details for
Quantity
  • Availability
  • Add to Cart