ENA428074387 Enamine

tert-butyl N-[(2-oxo-2,3-dihydro-1H-indol-3-yl)methyl]carbamate

  • MDL number MFCD11506127
  • Cas Number : 1086392-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181897
Availability details for
Quantity
  • Availability
  • Add to Cart