ENA428074390 Enamine

3-chloro-4-(piperazin-1-yl)benzonitrile

  • MDL number MFCD11038625
  • Cas Number : 182181-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-87
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181936
Availability details for
Quantity
  • Availability
  • Add to Cart