ENA428074402 Enamine

(5-tert-butyl-1,2,4-oxadiazol-3-yl)methanol

  • MDL number MFCD12138027
  • Cas Number : 1153452-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181957
Availability details for
Quantity
  • Availability
  • Add to Cart