ENA428074403 Enamine

3-amino-2-[(3,4,5-trifluorophenyl)methyl]propan-1-ol

  • MDL number MFCD17284551
  • Cas Number : 1310222-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181961
Availability details for
Quantity
  • Availability
  • Add to Cart