ENA428074409 Enamine

1-{4-[(6-aminopyridin-3-yl)methyl]piperazin-1-yl}-2-methylpropan-2-ol

  • MDL number MFCD20058341
  • Cas Number : 1484199-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181976
Availability details for
Quantity
  • Availability
  • Add to Cart