ENA428074412 Enamine

1-bromo-4-[(1S)-1-isocyanatoethyl]benzene

  • MDL number MFCD05664122
  • Cas Number : 149552-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-32
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182004
Availability details for
Quantity
  • Availability
  • Add to Cart