ENA428074420 Enamine

1-methyl-3,4-dihydroisoquinoline

  • MDL number MFCD00047648
  • Cas Number : 2412-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182056
Availability details for
Quantity
  • Availability
  • Add to Cart