ENA428074421 Enamine

1-cyclopentylethane-1,2-diamine

  • MDL number MFCD11503939
  • Cas Number : 1089358-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-182062
Availability details for
Quantity
  • Availability
  • Add to Cart