ENA428074430 Enamine

2-(2-ethylbutanamido)acetic acid

  • MDL number MFCD09811703
  • Cas Number : 98552-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182085
Availability details for
Quantity
  • Availability
  • Add to Cart