ENA428074431 Enamine

1-(4-bromonaphthalen-1-yl)-1-cyclopropylmethanamine

  • MDL number MFCD21960629
  • Cas Number : 1539832-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182104
Availability details for
Quantity
  • Availability
  • Add to Cart