ENA428074445 Enamine

3-bromo-4-(2-fluorophenyl)butan-2-one

  • MDL number MFCD16748496
  • Cas Number : 1250924-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182126
Availability details for
Quantity
  • Availability
  • Add to Cart