ENA428074448 Enamine

4-(2,2-difluoroethoxy)-3-(trifluoromethyl)aniline

  • MDL number MFCD14656334
  • Cas Number : 924909-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182130
Availability details for
Quantity
  • Availability
  • Add to Cart