ENA428074456 Enamine

4-(chloromethyl)-2-ethoxy-1-nitrobenzene

  • MDL number MFCD16699778
  • Cas Number : 1249505-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178978
Availability details for
Quantity
  • Availability
  • Add to Cart