ENA428074468 Enamine

2-cyano-3-cyclopropylprop-2-enoic acid

  • Cas Number : 98895-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182213
Availability details for
Quantity
  • Availability
  • Add to Cart