ENA428074470 Enamine

4-hydroxy-2-methoxybenzonitrile

  • MDL number MFCD08692760
  • Cas Number : 84224-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-157
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182223
Availability details for
Quantity
  • Availability
  • Add to Cart