ENA428074479 Enamine

3-nitro-2-(trifluoromethyl)phenol

  • MDL number MFCD16606322
  • Cas Number : 731002-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178985
Availability details for
Quantity
  • Availability
  • Add to Cart