ENA428074482 Enamine

2-bromo-3-chlorobenzene-1-sulfonamide

  • MDL number MFCD18390303
  • Cas Number : 1261499-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178987
Availability details for
Quantity
  • Availability
  • Add to Cart