ENA428074484 Enamine

2-methyl-1,3-thiazole-5-sulfonamide

  • MDL number MFCD18822170
  • Cas Number : 519055-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178988
Availability details for
Quantity
  • Availability
  • Add to Cart