ENA428074487 Enamine

[(2-fluoro-5-methylphenyl)methyl](methyl)amine

  • MDL number MFCD12824253
  • Cas Number : 1178036-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182309
Availability details for
Quantity
  • Availability
  • Add to Cart