ENA428074493 Enamine

tert-butyl N-(6-chloro-3-formylpyridin-2-yl)carbamate

  • MDL number MFCD10697513
  • Cas Number : 294659-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-178998
Availability details for
Quantity
  • Availability
  • Add to Cart