ENA428074502 Enamine

(2S,3S)-2-amino-N,N,3-trimethylpentanamide

  • MDL number MFCD14697078
  • Cas Number : 192821-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182408
Availability details for
Quantity
  • Availability
  • Add to Cart