ENA428074503 Enamine

4-(methylsulfanyl)benzene-1,2-diamine

  • MDL number MFCD18804703
  • Cas Number : 67469-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182409
Availability details for
Quantity
  • Availability
  • Add to Cart