ENA428074504 Enamine

6-amino-N-(pyrimidin-2-yl)pyridine-3-sulfonamide

  • MDL number MFCD17939132
  • Cas Number : 1281520-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182410
Availability details for
Quantity
  • Availability
  • Add to Cart