ENA428074505 Enamine

6-ethyl-7-hydroxy-3-(4-phenyl-4H-1,2,4-triazol-3-yl)-4H-chromen-4-one

  • MDL number MFCD07313696
  • Cas Number : 847758-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182411
Availability details for
Quantity
  • Availability
  • Add to Cart