ENA428074506 Enamine

3-bromo-4-(2,4-difluorophenyl)butan-2-one

  • MDL number MFCD16748461
  • Cas Number : 1247192-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182414
Availability details for
Quantity
  • Availability
  • Add to Cart