ENA428074508 Enamine

2-bromo-3-methoxycyclopent-2-en-1-one

  • MDL number MFCD11044009
  • Cas Number : 14203-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-120
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182417
Availability details for
Quantity
  • Availability
  • Add to Cart