ENA428074512 Enamine

4-cyano-3-methylbenzene-1-sulfonamide

  • MDL number MFCD18394315
  • Cas Number : 859490-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182429
Availability details for
Quantity
  • Availability
  • Add to Cart