ENA428074513 Enamine

2-(2-oxopropyl)-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD16164315
  • Cas Number : 1248216-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179008
Availability details for
Quantity
  • Availability
  • Add to Cart