ENA428074520 Enamine

2-({[(1R)-1-phenylethyl]amino}methyl)-3,4-dihydroquinazolin-4-one

  • MDL number MFCD28063921
  • Cas Number : 1406392-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179031
Availability details for
Quantity
  • Availability
  • Add to Cart