ENA428074521 Enamine

N-(4-chlorophenyl)prop-2-enamide

  • MDL number MFCD00045201
  • Cas Number : 5453-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179032
Availability details for
Quantity
  • Availability
  • Add to Cart