ENA428074524 Enamine

[4-(propan-2-yl)phenyl]methanesulfonamide

  • MDL number MFCD11212978
  • Cas Number : 64732-36-1
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-178392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178392
Availability details for
Quantity
  • Availability
  • Add to Cart