ENA428074526 Enamine

2-methyl-6-[3-(trifluoromethyl)phenyl]pyridine

  • MDL number MFCD18422115
  • Cas Number : 1261758-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178521
Availability details for
Quantity
  • Availability
  • Add to Cart