ENA428074529 Enamine

1-(3-chloro-4-fluorophenyl)-2,2,2-trifluoroethan-1-amine hydrochloride

  • MDL number MFCD28064098
  • Cas Number : 1798725-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182525
Availability details for
Quantity
  • Availability
  • Add to Cart