ENA428074533 Enamine

3-(3-hydroxyphenyl)-1-phenylurea

  • MDL number MFCD00446264
  • Cas Number : 13142-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-186
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182535
Availability details for
Quantity
  • Availability
  • Add to Cart