ENA428074534 Enamine

3-(4-hydroxyphenyl)-1-phenylurea

  • MDL number MFCD02668190
  • Cas Number : 2298-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182539
Availability details for
Quantity
  • Availability
  • Add to Cart