ENA428074535 Enamine

1,1,1-trifluoro-3-methylpentan-2-amine hydrochloride

  • MDL number MFCD28064099
  • Cas Number : 1787896-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182541
Availability details for
Quantity
  • Availability
  • Add to Cart