ENA428074541 Enamine

1-(4-amino-3-propoxyphenyl)piperidin-4-ol

  • MDL number MFCD18605330
  • Cas Number : 1411704-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182560
Availability details for
Quantity
  • Availability
  • Add to Cart