ENA428074543 Enamine

1-(2-bromoethyl)-2-ethylbenzene

  • MDL number MFCD20432717
  • Cas Number : 812631-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182563
Availability details for
Quantity
  • Availability
  • Add to Cart