ENA428074544 Enamine

2-cyclohexyl-4-methylpyridine

  • MDL number MFCD24539278
  • Cas Number : 15787-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182575
Availability details for
Quantity
  • Availability
  • Add to Cart