ENA428074545 Enamine

2-amino-3-[(2-methylprop-1-en-1-yl)sulfanyl]propanoic acid

  • MDL number MFCD24516145

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182576
Availability details for
Quantity
  • Availability
  • Add to Cart