ENA428074758 Enamine

tert-butyl N-{3-oxo-2-[(pyridin-3-yl)methyl]propyl}carbamate

  • MDL number MFCD28063805
  • Cas Number : 1798717-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183762
Availability details for
Quantity
  • Availability
  • Add to Cart