ENA428074850 Enamine

1-(3-aminocyclohexyl)ethan-1-one hydrochloride

  • MDL number MFCD28064143
  • Cas Number : 1955493-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-183925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183925
Availability details for
Quantity
  • Availability
  • Add to Cart